Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000596.2 | NP_000587.1 | 4 | 1464 | ||
GenBank mRNA | BC035263.1 | - | 4 | 1325 | 4800940 | |
BC057806.1 | - | 4 | 1336 | 30337849 | ||
M20841.1 | - | 4 | 1289 | |||
M31145.1 | - | 4 | 1335 | |||
X12385.1 | - | 4 | 1321 | |||
X13405.1 | - | 4 | 1273 | |||
X15002.1 | - | 3 | 751 | |||
Y00856.1 | - | 4 | 1221 |