Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161528.1 | NP_001155000.1 | 2, 3 | 2194 | ||
GenBank mRNA | BC026112.1 | - | 2, 3 | 730 | ||
BC144674.1 | - | 2, 3 | 289 | 9053208 | ||
BC144678.1 | - | 2, 3 | 312 | 9053212 |