Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC2HC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016010.2 | NP_057094.2 | 4 | 366 | ||
XM_005251256.1 | XP_005251313.1 | 4 | 366 | |||
XM_006716454.2 | XP_006716517.1 | 4 | 366 | |||
XM_011517539.2 | XP_011515841.1 | 4 | 366 | |||
GenBank mRNA | AF151820.1 | - | 4 | 318 | ||
AK024711.1 | - | 4 | 324 | |||
AK313723.1 | - | 4 | 324 | |||
BC009074.1 | - | 4 | 288 | 3868425 | ||
DA137885.1 | - | 4 | 366 |