Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016406.3 | NP_057490.2 | 5 | 610 | ||
XM_017001450.1 | XP_016856939.1 | 6 | 455 | |||
XM_017001451.1 | XP_016856940.1 | 6 | 719 | |||
GenBank mRNA | AB154405.1 | - | 5 | 356 | ||
AF151884.1 | - | 5 | 577 | |||
AF161504.1 | - | 5 | 596 | |||
AK292776.1 | - | 5 | 562 | |||
BC005187.1 | - | 5 | 602 | 3961569 | ||
CF128121.1 | - | 5 | 609 |