Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136157.1 | NP_001129629.1 | 1, 2 | 664 | ||
NM_001136158.1 | NP_001129630.1 | 1, 2 | 664 | |||
NM_001136159.1 | NP_001129631.1 | 1, 2 | 164 | |||
NM_017602.3 | NP_060072.1 | 1, 2 | 664 | |||
XM_005272622.3 | XP_005272679.1 | 1, 2 | 1414 | |||
XM_005278039.3 | XP_005278096.1 | 1, 2 | 1414 | |||
XM_005278042.4 | XP_005278099.1 | 1, 2 | 159 | |||
XM_006724537.2 | XP_006724600.1 | 1, 2 | 1414 | |||
XM_011543932.1 | XP_011542234.1 | 1, 2 | 1414 | |||
XM_017029628.1 | XP_016885117.1 | 1, 2 | 1414 | |||
XM_017029629.1 | XP_016885118.1 | 2, 3 | 158 | |||
XM_017029630.1 | XP_016885119.1 | 1, 2 | 159 | |||
GenBank mRNA | AK026260.1 | - | 1, 2 | 78 | ||
AK294590.1 | - | 1, 2 | 116 | |||
AL137509.1 | - | 1, 2 | 186 | |||
BC009917.2 | - | 1, 2 | 664 | 2958229 | ||
BC011738.2 | - | 1, 2 | 220 | |||
BC028225.1 | - | 1, 2 | 616 | 5241560 | ||
BC098440.1 | - | 2, 3 | 855 | 4840926 | ||
DC419901.1 | - | 1, 2 | 166 |