Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178817.3 | NP_848932.1 | 3 | 282 | ||
NM_206898.1 | NP_996781.1 | 3 | 282 | |||
XM_006724028.3 | XP_006724091.1 | 1 | 212 | |||
XM_017028407.1 | XP_016883896.1 | 1 | 208 | |||
GenBank mRNA | AF454915.1 | - | 3 | 282 | ||
AF483549.1 | - | 1 | 95 | |||
AY079152.1 | - | 1 | 95 | |||
BC062721.1 | - | 1 | 220 | 30327887 |