Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145079.1 | NP_001138551.1 | 4 | 492 | ||
NM_020751.2 | NP_065802.1 | 4 | 492 | |||
NR_026745.1 | 5 | 557 | ||||
XM_011535168.1 | XP_011533470.1 | 4 | 442 | |||
GenBank mRNA | AB032960.1 | - | 4 | 382 | ||
AK026638.1 | - | 3 | 159 | |||
AK294443.1 | - | 4 | 492 | |||
BC051723.1 | - | 4 | 442 | 5263056 | ||
CR627406.1 | - | 4 | 435 | |||
DC317375.1 | - | 4 | 492 |