Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007.4 | NP_000998.1 | 5 | 566 | ||
GenBank mRNA | AK097914.1 | - | 2367 | |||
AK222479.1 | - | 5 | 495 | |||
AK311745.1 | - | 5 | 556 | |||
BC000472.2 | - | 4 | 478 | 2961540 | ||
BC007308.1 | - | 4 | 410 | |||
BC071662.1 | - | 4 | 507 | 5505465 | ||
BC100903.1 | - | 4 | 470 | 40003600 | ||
BC100904.1 | - | 4 | 470 | 40003601 | ||
BU192706.1 | - | 5 | 608 | |||
CR456735.1 | - | 4 | 470 | |||
M22146.1 | - | 4 | 475 | |||
M58458.1 | - | 5 | 505 |