Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 196 | ||
XM_006716463.2 | XP_006716526.1 | 3 | 789 | |||
GenBank mRNA | AK225246.1 | - | 1 | 195 | ||
AK225254.1 | - | 1 | 194 | |||
AK291119.1 | - | 3 | 194 | |||
AK313365.1 | - | 3 | 194 | |||
BC006095.1 | - | 1 | 184 | 2821364 | ||
BC008850.2 | - | 1 | 221 | 4138113 | ||
BC009599.1 | - | 1 | 196 | 4134557 | ||
BC071671.1 | - | 1 | 177 | 4392472 | ||
BC071894.1 | - | 1 | 181 | 2906310 | ||
BC071895.1 | - | 1 | 179 | 6568654 | ||
BC087837.1 | - | 1 | 180 | 6170371 | ||
CR456773.1 | - | 1 | 174 | |||
L16558.1 | - | 1 | 174 | |||
X52967.1 | - | 3 | 195 | |||
X57958.1 | - | 1 | 211 | |||
X57959.1 | - | 1 | 184 |