Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105374297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136186.1 | 3 | 511 | |||
GenBank mRNA | BC044310.1 | - | 2, 3 | 206 | ||
DB075099.1 | - | 3 | 478 |