Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS1 SIRNA Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
UniGene ID:
Hs.485729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22809
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168407.1
NP_001161879.1
15
1925
NM_001168408.1
NP_001161880.1
15
1928
NM_001168409.1
NP_001161881.1
15
1713
NM_001168410.1
NP_001161882.1
15
2000
NM_014989.5
NP_055804.2
22
3707
XM_005248688.3
XP_005248745.1
20
3451
XM_011535604.2
XP_011533906.2
20
3448
XM_011535615.2
XP_011533917.2
20
3448
XM_017010517.1
XP_016866006.1
20
3451
XM_017010518.1
XP_016866007.1
20
3448
XM_017010520.1
XP_016866009.1
20
3451
XM_017010522.1
XP_016866011.1
20
3451
XM_017010523.1
XP_016866012.1
20
3451
XM_017010524.1
XP_016866013.1
20
3448
XM_017010527.1
XP_016866016.1
20
3448
XM_017010530.1
XP_016866019.1
20
3451
XM_017010531.1
XP_016866020.1
20
3448
XM_017010532.1
XP_016866021.1
20
3451
XM_017010534.1
XP_016866023.1
15
2024
XM_017010535.1
XP_016866024.1
15
2021
XM_017010537.1
XP_016866026.1
15
2021
XM_017010538.1
XP_016866027.1
15
2129
XM_017010539.1
XP_016866028.1
15
2016
XM_017010540.1
XP_016866029.1
15
2015
XM_017010541.1
XP_016866030.1
15
2012
XM_017010542.1
XP_016866031.1
15
2015
XM_017010543.1
XP_016866032.1
15
1931
XM_017010546.1
XP_016866035.1
15
2127
XM_017010547.1
XP_016866036.1
15
2124
XM_017010550.1
XP_016866039.1
15
2395
XM_017010551.1
XP_016866040.1
15
2392
XM_017010552.1
XP_016866041.1
15
2252
XM_017010553.1
XP_016866042.1
15
2249
XM_017010554.1
XP_016866043.1
15
2245
XM_017010557.1
XP_016866046.1
3
251
XM_017010558.1
XP_016866047.1
2
144
GenBank mRNA
AB051866.1
-
21
3481
AF263305.1
-
9
947
AF263306.1
-
8
869
AF263308.1
-
8
869
AF263309.1
-
8
866
AK293828.1
-
15
1603
AK294868.1
-
15
1925
AK295001.1
-
15
1846
AK296303.1
-
15
1769
AK308658.1
-
15
2000
AK309185.1
-
15
1713
AK316108.1
-
15
1726
AY190519.1
-
22
3386
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_052829, XM_008766936, AF199333, AF007836.This siRNA also targets the following transcripts in Mouse: NM_053270, NM_001012623, NM_183018, NM_001012624, AY356535, and others.
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline