Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024038.2 | NP_076943.1 | 3 | 894 | ||
GenBank mRNA | AF061732.1 | - | 2 | 469 | ||
AK027588.1 | - | 3 | 789 | |||
BC000216.2 | - | 3 | 894 | 2961319 | ||
BC004533.2 | - | 3 | 839 | 3139656 | ||
BC015347.1 | - | 3 | 853 | 4156774 |