Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093755.1 | NP_001087224.1 | 13 | 1516 | ||
NM_001093756.1 | NP_001087225.1 | 12 | 1495 | |||
NM_001243737.1 | NP_001230666.1 | 12 | 1498 | |||
NM_024941.3 | NP_079217.2 | 13 | 1513 | |||
XM_011543652.2 | XP_011541954.1 | 11 | 1099 | |||
XM_017009890.1 | XP_016865379.1 | 11 | 1364 | |||
GenBank mRNA | AK023673.1 | - | 13 | 1200 | ||
AL713699.1 | - | 5 | 554 | |||
BC047475.1 | - | 2 | 1321 | |||
BC117128.1 | - | 11 | 1132 | |||
BC142994.1 | - | 12 | 1185 | 8860449 | ||
BC143691.1 | - | 11 | 1135 | |||
BC143693.1 | - | 11 | 1132 | |||
BX641157.1 | - | 10 | 4464 | |||
DQ088673.1 | - | 11 | 997 |