Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190467.1 | NP_001177396.1 | 4 | 585 | ||
NR_104345.1 | 4 | 585 | ||||
GenBank mRNA | AL537525.3 | - | 4 | 521 | ||
BE791180.1 | - | 3 | 329 |