Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228345
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
7
481
NM_032291.3
NP_115667.2
7
739
XM_005271264.3
XP_005271321.1
7
573
XM_005271268.3
XP_005271325.1
6
489
XM_005271270.4
XP_005271327.1
7
820
XM_006710961.2
XP_006711024.1
7
573
XM_006710966.2
XP_006711029.1
7
573
XM_006710967.2
XP_006711030.1
7
561
XM_006710969.2
XP_006711032.1
6
489
XM_006710971.2
XP_006711034.1
7
573
XM_006710972.2
XP_006711035.1
7
573
XM_006710973.2
XP_006711036.1
7
573
XM_006710974.2
XP_006711037.1
6
489
XM_011542291.1
XP_011540593.1
7
736
XM_011542292.1
XP_011540594.1
7
736
XM_011542293.1
XP_011540595.1
6
489
XM_017002505.1
XP_016857994.1
7
561
XM_017002506.1
XP_016857995.1
8
684
XM_017002507.1
XP_016857996.1
7
736
XM_017002508.1
XP_016857997.1
8
682
XM_017002509.1
XP_016857998.1
7
561
XM_017002510.1
XP_016857999.1
8
684
XM_017002511.1
XP_016858000.1
8
684
XM_017002512.1
XP_016858001.1
8
557
XM_017002513.1
XP_016858002.1
7
736
XM_017002514.1
XP_016858003.1
6
489
XM_017002515.1
XP_016858004.1
6
489
XM_017002516.1
XP_016858005.1
7
612
XM_017002517.1
XP_016858006.1
8
683
XM_017002518.1
XP_016858007.1
7
611
XM_017002519.1
XP_016858008.1
7
636
XM_017002520.1
XP_016858009.1
6
489
XM_017002521.1
XP_016858010.1
6
489
XM_017002522.1
XP_016858011.1
7
611
XM_017002523.1
XP_016858012.1
7
635
XM_017002524.1
XP_016858013.1
8
677
XM_017002525.1
XP_016858014.1
8
676
XM_017002526.1
XP_016858015.1
7
561
XM_017002527.1
XP_016858016.1
8
585
XM_017002528.1
XP_016858017.1
7
605
XM_017002529.1
XP_016858018.1
7
573
XM_017002530.1
XP_016858019.1
7
561
XM_017002531.1
XP_016858020.1
7
573
XM_017002532.1
XP_016858021.1
6
489
XM_017002533.1
XP_016858022.1
8
675
XM_017002534.1
XP_016858023.1
6
489
XM_017002535.1
XP_016858024.1
7
603
XM_017002536.1
XP_016858025.1
6
489
XM_017002537.1
XP_016858026.1
7
602
GenBank mRNA
AB210039.1
-
7
521
AK090697.1
-
1
38
AK299941.1
-
6
500
AL136561.1
-
7
413
BC040516.1
-
8
486
5275011
BX640813.1
-
7
458
CR749541.1
-
7
675
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0