Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032340.3 | NP_115716.1 | 3, 4 | 357 | ||
GenBank mRNA | AF086234.1 | - | 3, 4 | 279 | ||
AK127468.1 | - | 1 | 916 | |||
AK311835.1 | - | 3, 4 | 306 | |||
AY568085.1 | - | 3, 4 | 333 | |||
BC006007.1 | - | 3, 4 | 333 | 4284544 | ||
BI604191.1 | - | 3, 4 | 307 | |||
DA123098.1 | - | 3, 4 | 357 |