Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172309.1 | NP_001165780.1 | 4, 5 | 550 | ||
NM_144573.3 | NP_653174.3 | 5, 6 | 742 | |||
XM_005271322.3 | XP_005271379.1 | 5, 6 | 633 | |||
XM_005271324.4 | XP_005271381.1 | 4, 5 | 537 | |||
XM_005271325.3 | XP_005271382.1 | 5, 6 | 633 | |||
GenBank mRNA | AF114264.1 | - | 1, 2 | 172 | ||
AF176780.1 | - | 4, 5 | 421 | |||
AK057954.1 | - | 1, 2 | 150 | |||
AK298565.1 | - | 4, 5 | 519 | |||
BC114444.1 | - | 1, 2 | 148 | 40083839 | ||
BC114445.1 | - | 1, 2 | 148 | 40083847 | ||
BX648546.1 | - | 5, 6 | 630 | |||
DC412178.1 | - | 551 | ||||
DQ464902.1 | - | 5, 6 | 629 | |||
S67069.1 | - | 1, 2 | 124 |