Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286829.1 | NP_001273758.1 | 9 | 1156 | ||
NM_145201.5 | NP_660202.3 | 9 | 1156 | |||
XM_017013975.1 | XP_016869464.1 | 8 | 2718 | |||
XM_017013976.1 | XP_016869465.1 | 8 | 2712 | |||
XM_017013978.1 | XP_016869467.1 | 8 | 2718 | |||
XM_017013979.1 | XP_016869468.1 | 8 | 914 | |||
XM_017013980.1 | XP_016869469.1 | 8 | 1019 | |||
GenBank mRNA | AF258565.1 | - | 3 | 604 | ||
AK097002.1 | - | 9 | 1148 | |||
AY214325.1 | - | 9 | 1144 | |||
AY214326.1 | - | 9 | 1144 | |||
AY214327.1 | - | 9 | 1144 | |||
BC006284.2 | - | 9 | 1128 | 3957135 | ||
BC014190.2 | - | 9 | 1128 | |||
BC032466.1 | - | 4 | 442 | 5210589 |