Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276252.1 | NP_001263181.1 | 14 | 2103 | ||
NM_015023.4 | NP_055838.2 | 14 | 2100 | |||
XM_011541057.1 | XP_011539359.1 | 14 | 1781 | |||
GenBank mRNA | AB028960.1 | - | 9 | 1242 | ||
AK001734.1 | - | 13 | 1575 | |||
AK023101.1 | - | 5 | 642 | |||
AK023778.1 | - | 5 | 682 | |||
AK315564.1 | - | 14 | 1987 | |||
BC032523.1 | - | 14 | 1963 | 5500384 |