Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129778.1 | NP_001123250.1 | 3, 4 | 454 | ||
XM_017024637.1 | XP_016880126.1 | 4, 5 | 533 | |||
GenBank mRNA | BC157869.1 | - | 3, 4 | 454 | 9057088 |