Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139514.1 | NP_001132986.1 | 7 | 1505 | ||
NM_001139515.1 | NP_001132987.1 | 8 | 930 | |||
NM_053281.3 | NP_444511.1 | 8 | 1544 | |||
XM_011530846.2 | XP_011529148.1 | 8 | 1543 | |||
XM_011530847.2 | XP_011529149.1 | 8 | 1543 | |||
XM_011530848.2 | XP_011529150.1 | 8 | 1543 | |||
XM_017029254.1 | XP_016884743.1 | 7 | 1504 | |||
XM_017029255.1 | XP_016884744.1 | 7 | 1504 | |||
XM_017029256.1 | XP_016884745.1 | 7 | 1504 | |||
GenBank mRNA | AF428101.1 | - | 8 | 1374 | ||
AK055953.1 | - | 7 | 891 | |||
AK091842.1 | - | 7 | 1498 | |||
AK290596.1 | - | 8 | 1544 | |||
AK294498.1 | - | 3 | 339 | |||
AK295648.1 | - | 3 | 340 | |||
AK298787.1 | - | 8 | 930 | |||
AL834456.1 | - | 1 | 136 | |||
BC114950.1 | - | 8 | 1453 | 40006598 |