Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 13 | 2801 | ||
XM_006712654.2 | XP_006712717.1 | 13 | 3378 | |||
XM_006712655.3 | XP_006712718.1 | 9 | 1015 | |||
XM_011511530.2 | XP_011509832.1 | 12 | 2513 | |||
XM_017004569.1 | XP_016860058.1 | 12 | 2505 | |||
XM_017004570.1 | XP_016860059.1 | 12 | 2505 | |||
XM_017004571.1 | XP_016860060.1 | 12 | 2443 | |||
XM_017004572.1 | XP_016860061.1 | 10 | 2093 | |||
GenBank mRNA | AB046783.2 | - | 12 | 2522 | ||
AB053305.1 | - | 13 | 2568 | |||
AF391100.1 | - | 13 | 2568 | |||
AK056413.1 | - | 4 | 406 | |||
AK291029.1 | - | 13 | 2585 | |||
BC071576.1 | - | 13 | 2563 | 30347073 |