Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM135A SIRNA Assays ›
Gene Symbol:
FAM135A
Gene Name:
family with sequence similarity 135 member A
Gene Aliases:
KIAA1411
Chromosome Location:
Chr.6: 70413404 - 70561174 on Build GRCh38
UniGene ID:
Hs.211700
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229568
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM135A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105531.2
NP_001099001.1
15
1804
NM_001162529.1
NP_001156001.1
13
2007
NM_020819.4
NP_065870.3
14
1667
XM_005248743.1
XP_005248800.1
16
2388
XM_005248744.4
XP_005248801.1
15
2355
XM_005248745.2
XP_005248802.1
15
2310
XM_005248748.2
XP_005248805.1
16
1800
XM_005248749.1
XP_005248806.1
16
2388
XM_005248751.2
XP_005248808.1
16
1800
XM_011535993.2
XP_011534295.1
15
2108
XM_011535994.2
XP_011534296.1
16
2528
XM_011535995.2
XP_011534297.1
16
2329
XM_011535996.2
XP_011534298.1
16
2486
XM_011535997.2
XP_011534299.1
14
2230
XM_017011124.1
XP_016866613.1
15
2361
XM_017011125.1
XP_016866614.1
14
2133
XM_017011126.1
XP_016866615.1
15
2447
XM_017011127.1
XP_016866616.1
14
2229
XM_017011128.1
XP_016866617.1
13
2153
XM_017011129.1
XP_016866618.1
16
1956
XM_017011130.1
XP_016866619.1
14
1603
XM_017011131.1
XP_016866620.1
15
1704
XM_017011132.1
XP_016866621.1
14
1691
XM_017011133.1
XP_016866622.1
13
1481
XM_017011134.1
XP_016866623.1
15
1722
XM_017011135.1
XP_016866624.1
14
2209
XM_017011136.1
XP_016866625.1
5
936
GenBank mRNA
AB037832.1
-
13
1975
AK128547.1
-
1
43
AL136820.1
-
2
632
BC131782.1
-
15
1630
40124841
CR749254.1
-
14
1505
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular lipid metabolic process
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0