Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000713.2 | NP_000704.1 | 5 | 830 | ||
GenBank mRNA | BC109371.1 | - | 5 | 747 | 5229026 | |
BU528444.1 | - | 3 | 425 | |||
D26308.1 | - | 5 | 778 | |||
D32143.1 | - | 5 | 808 | |||
EU794593.1 | - | 5 | 830 |