Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002752.4 | NP_002743.3 | 10, 11 | 1334 | ||
NM_139068.2 | NP_620707.1 | 10, 11 | 1334 | |||
NM_139069.2 | NP_620708.1 | 10, 11 | 1334 | |||
NM_139070.2 | NP_620709.1 | 10, 11 | 1334 | |||
XM_005265940.4 | XP_005265997.1 | 11, 12 | 1467 | |||
XM_006714891.2 | XP_006714954.1 | 11, 12 | 1392 | |||
XM_017009638.1 | XP_016865127.1 | 11, 12 | 1303 | |||
XM_017009639.1 | XP_016865128.1 | 12, 13 | 1560 | |||
XM_017009640.1 | XP_016865129.1 | 11, 12 | 1451 | |||
XM_017009641.1 | XP_016865130.1 | 11, 12 | 1401 | |||
XM_017009642.1 | XP_016865131.1 | 10, 11 | 1230 | |||
GenBank mRNA | AB451302.1 | - | 9, 10 | 1063 | ||
AB451355.1 | - | 9, 10 | 1063 | |||
AB451433.1 | - | 9, 10 | 1063 | |||
AK289638.1 | - | 10, 11 | 1230 | |||
BC032539.1 | - | 9, 10 | 1124 | 5528624 | ||
CR536580.1 | - | 9, 10 | 1063 | |||
HM036707.1 | - | 9, 10 | 1083 | |||
HM036708.1 | - | 9, 10 | 1083 | |||
HM036709.1 | - | 10, 11 | 1324 | |||
L31951.1 | - | 9, 10 | 1121 | |||
U09759.1 | - | 9, 10 | 1214 | |||
U34821.1 | - | 9, 10 | 1107 | |||
U35002.1 | - | 9, 10 | 1085 | |||
U35003.1 | - | 9, 10 | 1097 |