Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 3 | 299 | ||
NM_001115008.2 | NP_001108480.1 | 3 | 500 | |||
NM_001288997.1 | NP_001275926.1 | 3 | 237 | |||
NM_194282.3 | NP_919258.2 | 3 | 1163 | |||
NR_110254.1 | 3 | 500 | ||||
XM_005262750.4 | XP_005262807.1 | 3 | 972 | |||
XM_006714081.3 | XP_006714144.1 | 3 | 898 | |||
XM_017007729.1 | XP_016863218.1 | 3 | 1105 | |||
XM_017007730.1 | XP_016863219.1 | 3 | 494 | |||
GenBank mRNA | AB111889.1 | - | 3 | 487 | ||
AK292769.1 | - | 3 | 247 | |||
AK294256.1 | - | 3 | 1092 | |||
BX537919.1 | - | 3 | 397 | |||
BX640657.1 | - | 3 | 226 | |||
BX640966.1 | - | 3 | 1158 | |||
CX871759.1 | - | 2 | 770 |