Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006120.3 | NP_001006120.2 | 5, 6 | 704 | ||
NM_001320949.1 | NP_001307878.1 | 5, 6 | 704 | |||
NM_001320950.1 | NP_001307879.1 | 6, 7 | 755 | |||
XM_011531486.2 | XP_011529788.1 | 5, 6 | 577 | |||
XM_011531487.2 | XP_011529789.1 | 5, 6 | 664 | |||
XM_011531488.2 | XP_011529790.1 | 5, 6 | 664 | |||
XM_011531489.1 | XP_011529791.1 | 5, 6 | 664 | |||
XM_011531490.1 | XP_011529792.1 | 4, 5 | 503 | |||
GenBank mRNA | BC144422.1 | - | 5, 6 | 617 | 9052943 | |
BC144423.1 | - | 6, 7 | 668 |
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320944.1 | NP_001307873.1 | 5, 6 | 704 | ||
NM_001320945.1 | NP_001307874.1 | 4, 5 | 526 | |||
NM_005058.3 | NP_005049.1 | 5, 6 | 704 | |||
XM_011531437.2 | XP_011529739.1 | 5, 6 | 643 | |||
XM_011531438.1 | XP_011529740.1 | 5, 6 | 643 | |||
XM_017030060.1 | XP_016885549.1 | 4, 5 | 480 | |||
GenBank mRNA | BC047768.1 | - | 4, 5 | 501 | 5744153 | |
BC070298.1 | - | 5, 6 | 669 | 6615329 | ||
BC146853.1 | - | 5, 6 | 617 | 9056781 | ||
X76059.1 | - | 5, 6 | 669 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006121.3 | NP_001006121.1 | 5, 6 | 704 | ||
NM_001320948.1 | NP_001307877.1 | 5, 6 | 704 | |||
XM_011531433.1 | XP_011529735.1 | 5, 6 | 664 | |||
XM_011531434.1 | XP_011529736.1 | 4, 5 | 503 | |||
XM_017030056.1 | XP_016885545.1 | 5, 6 | 664 | |||
XM_017030057.1 | XP_016885546.1 | 4, 5 | 480 | |||
GenBank mRNA | BC136862.1 | - | 5, 6 | 617 | 9020853 | |
BC140758.1 | - | 5, 6 | 617 | 9021626 | ||
BC144459.1 | - | 5, 6 | 617 | 9052985 |
Gene Symbol: RBMY1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006118.2 | NP_001006118.2 | 5, 6 | 664 | ||
XM_011531493.1 | XP_011529795.1 | 5, 6 | 664 | |||
XM_011531494.2 | XP_011529796.1 | 5, 6 | 664 | |||
XM_011531495.1 | XP_011529797.1 | 5, 6 | 664 | |||
XM_011531496.1 | XP_011529798.1 | 4, 5 | 503 | |||
XM_017030058.1 | XP_016885547.1 | 4, 5 | 480 |
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 5, 6 | 664 | ||
NM_152585.2 | NP_689798.1 | 5, 6 | 664 | |||
XM_017030032.1 | XP_016885521.1 | 2, 3 | 253 | |||
XM_017030033.1 | XP_016885522.1 | 4, 5 | 480 | |||
GenBank mRNA | BC030018.2 | - | 5, 6 | 664 | 5269012 | |
X76060.1 | - | 5, 6 | 646 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 5, 6 | 729 | ||
XM_011531500.1 | XP_011529802.1 | 5, 6 | 600 | |||
XM_011531502.1 | XP_011529804.1 | 5, 6 | 600 | |||
XM_011531503.1 | XP_011529805.1 | 2, 3 | 253 | |||
XM_017030059.1 | XP_016885548.1 | 4, 5 | 480 | |||
GenBank mRNA | AK313597.1 | - | 5, 6 | 733 | ||
BC144577.1 | - | 5, 6 | 617 | 9053106 | ||
BC152392.1 | - | 4, 5 | 574 | 8860211 |