Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 16 | 2630 | ||
XM_005268466.1 | XP_005268523.1 | 16 | 2683 | |||
XM_005268467.1 | XP_005268524.1 | 15 | 2518 | |||
XM_005268468.1 | XP_005268525.1 | 15 | 2515 | |||
XM_006714787.1 | XP_006714850.1 | 17 | 2647 | |||
XM_011537661.1 | XP_011535963.1 | 15 | 2386 | |||
XM_011537662.1 | XP_011535964.1 | 15 | 2383 | |||
XM_017009587.1 | XP_016865076.1 | 17 | 2644 | |||
GenBank mRNA | AB037732.1 | - | 12 | 1953 | ||
AK128738.1 | - | 15 | 2515 | |||
AL833706.1 | - | 3 | 280 | |||
BC033524.1 | - | 4 | 473 | |||
BX647384.1 | - | 16 | 2630 |