Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302489.1 | NP_001289418.1 | 9 | 929 | ||
NM_001302490.1 | NP_001289419.1 | 8 | 861 | |||
NM_001302491.1 | NP_001289420.1 | 7 | 768 | |||
NM_001302492.1 | NP_001289421.1 | 8 | 886 | |||
NM_001610.3 | NP_001601.1 | 9 | 957 | |||
GenBank mRNA | AK298122.1 | - | 7 | 723 | ||
AK298439.1 | - | 9 | 932 | |||
AK299382.1 | - | 9 | 886 | |||
AK300569.1 | - | 8 | 885 | |||
AK300911.1 | - | 8 | 858 | |||
AK301860.1 | - | 8 | 819 | |||
AK303894.1 | - | 8 | 853 | |||
AK308512.1 | - | 7 | 1176 | |||
AK311431.1 | - | 8 | 851 | |||
AY007137.1 | - | 6 | 489 | |||
BC003160.1 | - | 9 | 902 | 2957907 | ||
BC093010.1 | - | 9 | 915 | 30332133 | ||
X12548.1 | - | 9 | 911 |