Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152769.2 | NP_689982.3 | 9 | 2158 | ||
XM_017026559.1 | XP_016882048.1 | 9 | 2277 | |||
GenBank mRNA | CK824551.1 | - | 1 | 458 |