Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111298.2 | NP_001104768.2 | 3 | 986 | ||
NM_001286360.1 | NP_001273289.1 | 2 | 890 | |||
NM_001286361.1 | NP_001273290.1 | 2 | 890 | |||
NM_173672.4 | NP_775943.1 | 3 | 986 | |||
NR_104429.1 | 3 | 986 | ||||
XM_011535765.2 | XP_011534067.1 | 2 | 350 | |||
XM_011535766.2 | XP_011534068.1 | 3 | 454 | |||
XM_011535767.2 | XP_011534069.1 | 3 | 454 | |||
XM_011535769.2 | XP_011534071.1 | 3 | 454 | |||
XM_017010774.1 | XP_016866263.1 | 3 | 454 | |||
GenBank mRNA | AI052546.1 | - | 2 | 247 | ||
AK293674.1 | - | 3 | 438 | |||
AK315790.1 | - | 3 | 459 | |||
BC038716.1 | - | 3 | 445 | 5268756 | ||
BI824055.1 | - | 2 | 223 | |||
DA413621.1 | - | 2 | 273 | |||
DC419072.1 | - | 3 | 459 | |||
DQ363562.1 | - | 3 | 986 | |||
DQ423529.1 | - | 3 | 986 |