Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229972
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
2364
NM_001174093.1
NP_001167564.1
6
2024
NM_001174094.1
NP_001167565.1
7
2361
NM_001174095.1
NP_001167566.1
6
1883
NM_001174096.1
NP_001167567.1
7
2087
NM_001323638.1
NP_001310567.1
11
2734
NM_001323641.1
NP_001310570.1
7
2314
NM_001323642.1
NP_001310571.1
9
2616
NM_001323643.1
NP_001310572.1
9
2394
NM_001323644.1
NP_001310573.1
6
2113
NM_001323645.1
NP_001310574.1
8
2457
NM_001323646.1
NP_001310575.1
10
2695
NM_001323647.1
NP_001310576.1
8
2227
NM_001323648.1
NP_001310577.1
7
2167
NM_001323649.1
NP_001310578.1
8
2230
NM_001323650.1
NP_001310579.1
10
2462
NM_001323651.1
NP_001310580.1
8
2130
NM_001323652.1
NP_001310581.1
9
2279
NM_001323653.1
NP_001310582.1
9
2386
NM_001323654.1
NP_001310583.1
9
2273
NM_001323655.1
NP_001310584.1
9
2383
NM_001323656.1
NP_001310585.1
7
2058
NM_001323657.1
NP_001310586.1
7
2061
NM_001323658.1
NP_001310587.1
7
1900
NM_001323659.1
NP_001310588.1
7
2118
NM_001323660.1
NP_001310589.1
7
2115
NM_001323661.1
NP_001310590.1
8
2261
NM_001323662.1
NP_001310591.1
9
2663
NM_001323663.1
NP_001310592.1
10
2602
NM_001323664.1
NP_001310593.1
9
2586
NM_001323665.1
NP_001310594.1
8
2504
NM_001323666.1
NP_001310595.1
7
2444
NM_001323671.1
NP_001310600.1
8
2340
NM_001323672.1
NP_001310601.1
9
2392
NM_001323673.1
NP_001310602.1
8
2337
NM_001323674.1
NP_001310603.1
5
1862
NM_001323675.1
NP_001310604.1
5
1972
NM_001323676.1
NP_001310605.1
7
2197
NM_001323677.1
NP_001310606.1
7
2194
NM_001323678.1
NP_001310607.1
5
2139
NM_030751.5
NP_110378.3
7
2084
XM_006717498.2
XP_006717561.1
10
2522
XM_011519643.2
XP_011517945.1
10
2825
XM_017016597.1
XP_016872086.1
10
2424
XM_017016598.1
XP_016872087.1
10
5554
XM_017016599.1
XP_016872088.1
9
5392
XM_017016600.1
XP_016872089.1
11
5633
XM_017016601.1
XP_016872090.1
10
5551
XM_017016602.1
XP_016872091.1
11
5630
XM_017016603.1
XP_016872092.1
10
5468
GenBank mRNA
AK091478.1
-
6
1984
AK294775.1
-
7
2279
AK296244.1
-
7
2007
AK299541.1
-
4
1621
AK299666.1
-
7
2118
AK300830.1
-
6
1831
AK307510.1
-
8
2090
AK314683.1
-
7
2045
AL831979.1
-
9
2342
BC009572.1
-
7
2040
BC112392.1
-
7
2040
40036600
BX647794.1
-
11
2735
U12170.1
-
7
2045
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0