Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134405.1 | NP_001127877.1 | 9, 10 | 1512 | ||
NM_138290.2 | NP_612147.1 | 10, 11 | 1563 | |||
XM_005250158.2 | XP_005250215.1 | 7, 8 | 987 | |||
XM_011515828.2 | XP_011514130.1 | 10, 11 | 1046 | |||
XM_011515829.2 | XP_011514131.1 | 9, 10 | 1180 | |||
XM_017011761.1 | XP_016867250.1 | 10, 11 | 1250 | |||
XM_017011762.1 | XP_016867251.1 | 11, 12 | 1175 | |||
XM_017011763.1 | XP_016867252.1 | 8, 9 | 989 | |||
GenBank mRNA | AK055233.1 | - | 10, 11 | 1561 | ||
AK294039.1 | - | 9, 10 | 1310 |