Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 13 | 3583 | ||
XM_005259311.4 | XP_005259368.1 | 14 | 4523 | |||
XM_005259314.3 | XP_005259371.1 | 12 | 2609 | |||
XM_011527367.2 | XP_011525669.1 | 14 | 4538 | |||
XM_011527368.2 | XP_011525670.1 | 15 | 3876 | |||
XM_011527369.2 | XP_011525671.1 | 15 | 3880 | |||
XM_011527370.2 | XP_011525672.1 | 14 | 3737 | |||
XM_011527371.2 | XP_011525673.1 | 14 | 3724 | |||
XM_011527372.2 | XP_011525674.1 | 12 | 4373 | |||
XM_011527373.2 | XP_011525675.1 | 13 | 4364 | |||
XM_011527374.2 | XP_011525676.1 | 14 | 4373 | |||
GenBank mRNA | AK314169.1 | - | 10 | 2142 | ||
AL136846.1 | - | 11 | 1952 | |||
AL833862.1 | - | 5 | 1580 | |||
BC101050.2 | - | 11 | 1949 | 40008070 | ||
BC101051.2 | - | 11 | 1949 | 40008071 | ||
BC101052.1 | - | 11 | 1949 | 40008072 | ||
BC101053.2 | - | 11 | 1949 | 40008073 |