Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM25G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137549.1 | NP_001131021.1 | 3 | 223 |
Gene Symbol: FAM25BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104039.1 | 3 | 223 | |||
GenBank mRNA | BC146968.1 | - | 3 | 220 | 9056896 |
Gene Symbol: FAM25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137548.2 | NP_001131020.1 | 3 | 223 | ||
GenBank mRNA | BC146955.1 | - | 3 | 220 | 9056883 | |
BC146972.1 | - | 3 | 223 | 9056900 | ||
BC146989.1 | - | 3 | 223 | 9056917 | ||
BC146990.1 | - | 3 | 223 | 9056918 | ||
BC146991.1 | - | 3 | 223 | 9056919 | ||
BC147022.1 | - | 3 | 223 | 9056950 | ||
BC147026.1 | - | 3 | 223 | 9056954 | ||
BC171871.1 | - | 3 | 223 | 9054525 | ||
BX093303.1 | - | 3 | 278 |