Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST3H2BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175055.2 | NP_778225.1 | 1 | 66 | ||
GenBank mRNA | AK091220.1 | - | 1 | 66 | ||
BC100855.1 | - | 1 | 65 | 40002412 | ||
BC100856.1 | - | 1 | 65 | 40002414 | ||
BC100857.1 | - | 1 | 65 | 40002416 | ||
BC100858.1 | - | 1 | 65 | 40002418 | ||
BC137467.1 | - | 1 | 65 | 9021467 | ||
BC137468.1 | - | 1 | 65 | 9021468 |