Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 1 | 112 | ||
NM_181268.2 | NP_851785.2 | 1 | 112 | |||
NM_181269.2 | NP_851786.1 | 1 | 168 | |||
NM_181270.2 | NP_851787.1 | 1 | 168 | |||
NM_181271.2 | NP_851788.1 | 1 | 168 | |||
NM_181272.2 | NP_851789.1 | 1 | 168 | |||
NM_181283.2 | NP_851800.1 | 1 | 168 | |||
NM_181296.2 | NP_851813.1 | 1 | 168 | |||
GenBank mRNA | AY174118.1 | - | 1 | 172 | ||
AY174119.1 | - | 1 | 172 | |||
AY174120.1 | - | 1 | 172 | |||
AY174121.1 | - | 1 | 172 | |||
AY174122.1 | - | 1 | 172 | |||
AY174123.1 | - | 1 | 172 | |||
AY174124.1 | - | 1 | 172 | |||
AY174125.1 | - | 1 | 172 | |||
AY174126.1 | - | 1 | 172 | |||
AY174127.1 | - | 1 | 172 | |||
AY174128.1 | - | 1 | 172 | |||
AY174130.1 | - | 1 | 172 | |||
AY174131.1 | - | 1 | 172 | |||
AY174132.1 | - | 1 | 172 | |||
AY174133.1 | - | 1 | 172 | |||
BC057852.1 | - | 1 | 112 | 5273108 | ||
DN992496.1 | - | 1 | 314 |