Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027484.2 | 4 | 393 | |||
GenBank mRNA | EB386870.1 | - | 2 | 126 |
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000566.3 | NP_000557.1 | 4 | 393 | ||
XM_005244957.3 | XP_005245014.1 | 4 | 396 | |||
XM_005244958.4 | XP_005245015.1 | 2 | 134 | |||
GenBank mRNA | AK291451.1 | - | 4 | 376 | ||
AK291502.1 | - | 4 | 393 | |||
BC032634.1 | - | 4 | 355 | 5248549 | ||
BC152383.1 | - | 4 | 346 | 100013442 | ||
DC320241.1 | - | 4 | 393 | |||
DQ786309.1 | - | 2 | 70 | |||
L03418.1 | - | 4 | 343 | |||
X14355.1 | - | 4 | 398 | |||
X14356.1 | - | 4 | 379 |