Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002307.3 | NP_002298.1 | 2, 3 | 125 | ||
GenBank mRNA | BP225960.1 | - | 2, 3 | 125 | ||
CR456917.1 | - | 1, 2 | 100 | |||
CV575179.1 | - | 2, 3 | 113 |
Gene Symbol: LGALS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042507.3 | NP_001035972.1 | 2, 3 | 117 | ||
GenBank mRNA | BC042911.2 | - | 1, 2 | 93 | ||
BC061588.1 | - | 2, 3 | 116 | 6202380 | ||
BC073743.1 | - | 2, 3 | 116 | 5478866 | ||
L07769.1 | - | 2, 3 | 118 | |||
U06643.1 | - | 2, 3 | 129 |