Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181880.1 | NP_870996.1 | 2 | 458 | ||
GenBank mRNA | BC056508.1 | - | 2 | 458 | 6450744 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104154.2 | - | 2 | 415 | 40029402 |
Gene Symbol: VCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004679.2 | NP_004670.1 | 2 | 462 | ||
GenBank mRNA | AF000979.1 | - | 2 | 462 | ||
BC096729.1 | - | 2 | 415 | 40014422 |