Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK3 SIRNA Assays ›
Gene Symbol:
MARK3
Gene Name:
microtubule affinity regulating kinase 3
Gene Aliases:
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location:
Chr.14: 103385364 - 103503831 on Build GRCh38
UniGene ID:
Hs.35828
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230621
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128918.2
NP_001122390.2
6
1134
NM_001128919.2
NP_001122391.2
6
1134
NM_001128920.2
NP_001122392.2
6
1134
NM_001128921.2
NP_001122393.2
6
1134
NM_002376.6
NP_002367.5
6
1134
XM_005267641.4
XP_005267698.2
6
513
XM_005267642.4
XP_005267699.2
6
1134
XM_005267643.4
XP_005267700.2
6
1134
XM_006720146.3
XP_006720209.1
6
508
XM_011536769.2
XP_011535071.1
5
247
XM_017021291.1
XP_016876780.1
6
1134
XM_017021292.1
XP_016876781.1
6
1134
XM_017021293.1
XP_016876782.1
7
609
XM_017021294.1
XP_016876783.1
5
883
XM_017021295.1
XP_016876784.1
5
898
XM_017021296.1
XP_016876785.1
5
915
XM_017021297.1
XP_016876786.1
5
891
XM_017021298.1
XP_016876787.1
5
891
XM_017021299.1
XP_016876788.1
5
942
XM_017021300.1
XP_016876789.1
4
368
XM_017021301.1
XP_016876790.1
4
523
XM_017021302.1
XP_016876791.1
4
368
XM_017021303.1
XP_016876792.1
4
368
XM_017021304.1
XP_016876793.1
4
368
XM_017021305.1
XP_016876794.1
4
368
XM_017021308.1
XP_016876797.1
5
1085
GenBank mRNA
AF387637.1
-
6
486
AF465413.1
-
6
978
AK307448.1
-
6
513
BC024773.1
-
6
957
5138958
BX161395.1
-
6
1064
M80359.1
-
6
639
U64205.1
-
6
843
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
establishment of cell polarity
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0