Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFASC SIRNA Assays ›
Gene Symbol:
NFASC
Gene Name:
neurofascin
Gene Aliases:
NF, NRCAML
Chromosome Location:
Chr.1: 204828652 - 205022822 on Build GRCh38
UniGene ID:
Hs.13349
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23065
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFASC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005388.2
NP_001005388.2
10
1165
NM_001005389.1
NP_001005389.2
10
1165
NM_001160331.1
NP_001153803.1
8
960
NM_001160332.1
NP_001153804.1
10
1198
NM_001160333.1
NP_001153805.1
9
1147
NM_015090.3
NP_055905.2
10
1198
XM_005244989.3
XP_005245046.2
10
1278
XM_005244991.3
XP_005245048.2
10
1278
XM_005244992.4
XP_005245049.2
10
1278
XM_005244993.3
XP_005245050.2
9
1227
XM_005244997.3
XP_005245054.2
9
1227
XM_011509311.1
XP_011507613.1
9
978
XM_011509312.1
XP_011507614.1
10
1278
XM_011509313.1
XP_011507615.1
9
978
XM_011509314.1
XP_011507616.1
9
1227
XM_011509315.1
XP_011507617.1
8
927
XM_011509316.1
XP_011507618.1
11
1175
XM_011509317.1
XP_011507619.1
9
978
XM_011509318.2
XP_011507620.1
10
1200
XM_011509319.1
XP_011507621.1
10
1124
XM_011509320.2
XP_011507622.1
9
1149
XM_011509321.1
XP_011507623.1
9
978
XM_011509322.1
XP_011507624.1
9
978
XM_011509323.1
XP_011507625.1
9
978
XM_011509324.1
XP_011507626.1
9
978
XM_011509325.1
XP_011507627.1
9
978
XM_011509326.1
XP_011507628.1
9
978
XM_011509327.1
XP_011507629.1
9
978
XM_011509328.1
XP_011507630.1
8
927
XM_017000733.1
XP_016856222.1
10
1278
XM_017000734.1
XP_016856223.1
9
1227
XM_017000735.1
XP_016856224.1
9
978
XM_017000736.1
XP_016856225.1
9
978
XM_017000737.1
XP_016856226.1
9
978
XM_017000738.1
XP_016856227.1
10
1200
XM_017000739.1
XP_016856228.1
10
1200
XM_017000740.1
XP_016856229.1
10
1200
XM_017000741.1
XP_016856230.1
7
781
XM_017000742.1
XP_016856231.1
8
927
XM_017000743.1
XP_016856232.1
9
1149
GenBank mRNA
AB018299.3
-
9
1029
AB177861.1
-
3
263
AK090639.1
-
10
1124
AK097802.1
-
8
975
AK295291.1
-
9
1126
AK299262.1
-
9
1126
AK308686.1
-
10
1278
BC040674.1
-
11
1416
BC117674.2
-
9
1014
BC137013.1
-
8
935
9021006
BC144454.1
-
8
902
9052980
BC144455.1
-
9
953
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
axon guidance
peripheral nervous system development
transmission of nerve impulse
paranodal junction assembly
heterotypic cell-cell adhesion
myelination
clustering of voltage-gated sodium channels
synapse organization
protein localization to juxtaparanode region of axon
protein targeting to plasma membrane
protein binding
protein domain specific binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0