Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032435.2 | NP_115811.2 | 10 | 3236 | ||
GenBank mRNA | AB058707.1 | - | 4 | 1433 | ||
AJ311798.1 | - | 10 | 3236 | |||
AK309811.1 | - | 3 | 600 | |||
BC136648.1 | - | 10 | 3095 | 9020638 | ||
BC136649.1 | - | 10 | 3095 | 9020639 |