Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128931.1 | NP_001122403.1 | 3 | 768 | ||
NM_138386.2 | NP_612395.2 | 3 | 768 | |||
XM_011532410.2 | XP_011530712.1 | 3 | 4235 | |||
XM_011532411.2 | XP_011530713.1 | 4 | 842 | |||
GenBank mRNA | AK097352.1 | - | 3 | 678 | ||
AK308601.1 | - | 3 | 737 | |||
BC008207.1 | - | 3 | 768 | 3447276 |