Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP2 SIRNA Assays ›
Gene Symbol:
CLASP2
Gene Name:
cytoplasmic linker associated protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 33496245 - 33718356 on Build GRCh38
UniGene ID:
Hs.108614
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23083
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207044.1
NP_001193973.1
25
2932
NM_015097.2
NP_055912.2
31
3354
XM_006713040.1
XP_006713103.1
33
3611
XM_006713041.1
XP_006713104.1
33
3608
XM_006713042.1
XP_006713105.1
32
3587
XM_006713048.1
XP_006713111.1
32
3548
XM_006713049.1
XP_006713112.1
31
3533
XM_006713050.1
XP_006713113.1
31
3524
XM_006713052.1
XP_006713115.1
31
3494
XM_006713053.1
XP_006713116.1
30
3473
XM_011533515.1
XP_011531817.1
31
3569
XM_017005947.1
XP_016861436.1
32
3584
XM_017005948.1
XP_016861437.1
31
3581
XM_017005949.1
XP_016861438.1
31
3412
XM_017005950.1
XP_016861439.1
32
3560
XM_017005951.1
XP_016861440.1
32
3557
XM_017005952.1
XP_016861441.1
31
3557
XM_017005953.1
XP_016861442.1
31
3554
XM_017005954.1
XP_016861443.1
32
3545
XM_017005955.1
XP_016861444.1
31
3536
XM_017005956.1
XP_016861445.1
30
3533
XM_017005957.1
XP_016861446.1
30
3521
XM_017005958.1
XP_016861447.1
30
3494
XM_017005959.1
XP_016861448.1
30
3491
XM_017005960.1
XP_016861449.1
29
3482
XM_017005961.1
XP_016861450.1
29
3325
XM_017005962.1
XP_016861451.1
30
3470
XM_017005963.1
XP_016861452.1
29
3470
XM_017005964.1
XP_016861453.1
29
3467
XM_017005965.1
XP_016861454.1
29
3455
XM_017005966.1
XP_016861455.1
28
3431
XM_017005967.1
XP_016861456.1
28
3274
XM_017005968.1
XP_016861457.1
26
2688
XM_017005969.1
XP_016861458.1
26
2698
XM_017005970.1
XP_016861459.1
26
2947
XM_017005971.1
XP_016861460.1
25
2674
XM_017005972.1
XP_016861461.1
26
3275
XM_017005973.1
XP_016861462.1
25
2920
XM_017005974.1
XP_016861463.1
24
2917
XM_017005975.1
XP_016861464.1
25
2637
XM_017005976.1
XP_016861465.1
23
2632
XM_017005977.1
XP_016861466.1
25
2896
XM_017005978.1
XP_016861467.1
24
2613
XM_017005979.1
XP_016861468.1
24
2623
XM_017005980.1
XP_016861469.1
25
3224
XM_017005981.1
XP_016861470.1
23
2610
XM_017005982.1
XP_016861471.1
23
2620
XM_017005983.1
XP_016861472.1
24
2872
XM_017005984.1
XP_016861473.1
24
2869
XM_017005985.1
XP_016861474.1
23
3197
XM_017005986.1
XP_016861475.1
22
2830
GenBank mRNA
AB014527.2
-
25
2571
AK297322.1
-
25
2607
BC011530.1
-
11
1169
BC140778.1
-
31
3190
9021646
BX640725.1
-
7
726
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
microtubule organizing center organization
regulation of microtubule polymerization or depolymerization
regulation of microtubule-based process
microtubule anchoring
positive regulation of exocytosis
cell division
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
positive regulation of extracellular matrix disassembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0