Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPATCH8 SIRNA Assays ›
Gene Symbol:
GPATCH8
Gene Name:
G-patch domain containing 8
Gene Aliases:
GPATC8, KIAA0553
Chromosome Location:
Chr.17: 44395277 - 44503602 on Build GRCh38
UniGene ID:
Hs.463129
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23101
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPATCH8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002909.3
NP_001002909.1
4
452
NM_001304939.1
NP_001291868.1
3
377
NM_001304940.1
NP_001291869.1
6
593
NM_001304941.1
NP_001291870.1
5
470
NM_001304942.1
NP_001291871.1
3
379
NM_001304943.1
NP_001291872.1
1
333
XM_011524557.1
XP_011522859.1
4
439
XM_011524558.1
XP_011522860.1
5
258
XM_011524559.2
XP_011522861.1
6
336
XM_011524561.2
XP_011522863.1
2
97
XM_017024372.1
XP_016879861.1
7
381
XM_017024373.1
XP_016879862.1
4
1216
XM_017024374.1
XP_016879863.1
6
299
XM_017024375.1
XP_016879864.1
5
481
XM_017024376.1
XP_016879865.1
6
341
XM_017024377.1
XP_016879866.1
5
318
XM_017024378.1
XP_016879867.1
6
499
XM_017024379.1
XP_016879868.1
7
381
XM_017024380.1
XP_016879869.1
5
282
GenBank mRNA
AB011125.2
-
3
1542
AK025600.1
-
4
254
BC136629.1
-
5
269
9020619
BC144323.1
-
5
269
BM722954.1
-
1
495
CN388517.1
-
3
157
CN388519.1
-
3
167
DA736259.1
-
6
380
DB021837.1
-
4
439
HY048299.1
-
4
453
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008768227, XM_006247478, XM_001062937, XM_006247479, XM_008768226.This siRNA also targets the following transcripts in Mouse: NM_001159492, BC035255, BY136728, CV555545, CJ057626.
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline