Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164211.1 | NP_001157683.1 | 20 | 2448 | ||
NM_015116.2 | NP_055931.1 | 19 | 2343 | |||
GenBank mRNA | AB023233.1 | - | 19 | 2304 | ||
AK123654.1 | - | 1 | 1109 | |||
AY050632.1 | - | 19 | 2336 | |||
BC117472.1 | - | 20 | 2349 | 40126023 | ||
BC143883.1 | - | 20 | 2349 | 9052399 | ||
CR739467.1 | - | 1 | 154 |