Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048194.2 | NP_001041659.1 | 11 | 2227 | ||
NM_001048195.2 | NP_001041660.1 | 11 | 2185 | |||
NM_001048199.2 | NP_001041664.1 | 12 | 2309 | |||
NM_001269.4 | NP_001260.1 | 10 | 2134 | |||
NR_030725.1 | 13 | 2452 | ||||
NR_030726.1 | 13 | 2448 | ||||
GenBank mRNA | BC001589.1 | - | 9 | 2071 | ||
BC010067.2 | - | 9 | 2053 | 4122227 | ||
X12654.1 | - | 10 | 2231 |