Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FCHO1 SIRNA Assays ›
Gene Symbol:
FCHO1
Gene Name:
FCH domain only 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 17747718 - 17788568 on Build GRCh38
UniGene ID:
Hs.96485
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23143
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FCHO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161357.1
NP_001154829.1
7
545
NM_001161358.1
NP_001154830.1
6
455
NM_001161359.1
NP_001154831.1
5
385
NM_015122.2
NP_055937.1
7
549
XM_005259831.3
XP_005259888.1
6
630
XM_006722701.3
XP_006722764.1
6
553
XM_006722702.3
XP_006722765.1
6
630
XM_011527824.2
XP_011526126.1
7
545
XM_011527825.2
XP_011526127.1
7
534
XM_011527826.2
XP_011526128.1
7
517
XM_011527827.2
XP_011526129.1
5
352
XM_011527828.2
XP_011526130.1
6
448
XM_017026519.1
XP_016882008.1
7
615
XM_017026520.1
XP_016882009.1
7
598
XM_017026521.1
XP_016882010.1
6
549
XM_017026522.1
XP_016882011.1
7
545
XM_017026523.1
XP_016882012.1
7
615
XM_017026524.1
XP_016882013.1
6
630
XM_017026525.1
XP_016882014.1
6
428
XM_017026526.1
XP_016882015.1
7
545
XM_017026527.1
XP_016882016.1
6
553
XM_017026528.1
XP_016882017.1
7
615
GenBank mRNA
AB006628.1
-
4
319
AK291410.1
-
6
411
AK303623.1
-
3
247
AK309403.1
-
3
298
AK316458.1
-
5
356
BC028021.1
-
7
490
5218319
BC041130.1
-
7
499
5757146
BI756664.1
-
7
545
DB158605.1
-
7
550
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
clathrin coat assembly
clathrin-mediated endocytosis
protein binding
AP-2 adaptor complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline