Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007099.2 | NP_001007100.1 | 2 | 162 | ||
NM_001007100.2 | NP_001007101.1 | 2 | 153 | |||
NM_001007250.2 | NP_001007251.1 | 2 | 162 | |||
NM_001193599.1 | NP_001180528.1 | 12 | 1387 | |||
NM_001193600.1 | NP_001180529.1 | 12 | 1327 | |||
NM_001193617.1 | NP_001180546.1 | 12 | 1401 | |||
NM_002979.4 | NP_002970.2 | 13 | 1459 | |||
XM_005271103.4 | XP_005271160.1 | 13 | 1402 | |||
XM_017002046.1 | XP_016857535.1 | 3 | 727 | |||
GenBank mRNA | AB208789.1 | - | 14 | 4154 | ||
AK294631.1 | - | 12 | 1330 | |||
AK295214.1 | - | 12 | 1344 | |||
AK303400.1 | - | 2 | 123 | |||
AK312856.1 | - | 13 | 1410 | |||
BC005911.1 | - | 2 | 152 | |||
BX537618.1 | - | 2 | 127 | |||
BX537619.1 | - | 2 | 127 | |||
CA867670.1 | - | 2 | 119 | |||
DB459183.1 | - | 2 | 131 | |||
M55421.1 | - | 2 | 162 | |||
M75883.1 | - | 13 | 1312 | |||
M75884.1 | - | 5 | 598 | |||
S52450.1 | - | 2 | 162 |